missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Eps15 Alexa Fluor 700-conjugated Antibody, R&D Systems™
Description
EPS15 Monoclonal specifically detects EPS15 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).
Specifications
Specifications
| Antigen | EPS15 |
| Applications | Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 1261C |
| Conjugate | Alexa Fluor 700 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Accession No. | P42566 |
| Gene Alias | AF1P, AF-1P, ALL1 fused gene from chromosome 1, epidermal growth factor receptor pathway substrate 15, epidermal growth factor receptor substrate 15, MLLT5, Protein AF-1p, Protein Eps15 |
| Host Species | Rabbit |
| Show More |