missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human EPO (Erythropoietin) Recombinant Protein DFS Item

Recombinant Protein

Supplier:  Gibco™ PHC2054

Catalog No. PHC2054


Only null left
Explore available promotions

Description

Description

Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute in sterile, deionized water to a concentration of 12,000-120,000 IU/mL. Further dilutions can be made in aqueous buffers. For long term storage, EPO solutions should be supplemented with carrier protein, such as 0.1% HSA or BSA. Storage: This lyophilized preparation is stable at room temperature for three weeks, but should be stored desiccated below -18°C. Upon reconstitution, the preparation is stable for up to one week at 2°C to 8°C, but should be stored below -18°C for long term storage. Avoid repeated freeze/thaw cycles.

Erythropoietin (EPO) protein is a growth factor essential for the survival, proliferation, and differentiation of erythroid precursors. EPO is not responsible for early commitment to the lineage, and instead induces the final maturation of partially differentiated erythroid cells and the production of red cell-specific proteins, such as hemoglobin. The EPO receptor (EPOR) is present in higher levels on precursor cells, and expression diminishes with cell maturation.Hepatocytes are the main source of EPO in the fetus, followed by interstitial cells of the kidney after birth and through adulthood. EPO expression is upregulated during hypoxia, and low levels of transcription have also been detected in lung and brain tissue under these conditions. Recent data suggest that EPO may also play a role in the protection of neuronal tissue from damage and initiation of repair.
Specifications

Specifications

P01588
0.006mg citric acid, 0.59mg sodium citrate with 0.58mg NaCl and no preservative
30.4 kDa
Purified
-20°C
<0.1 ng/μg
Epo
Specific Activity = 150,000 IU/mg; determined by measuring normocythaemic mice.
Unconjugated
Recombinant
-20°C
Specific Activity = 150,000 IU/mg; determined by measuring normocythaemic mice.
Approved for shipment on Wet or Dry Ice
Growth Factors & Receptors
Lyophilized
Neurobiology, Stem Cell Research, Differentiation
>98% by SDS-PAGE
Bioactivity
2056
Human EPO (Erythropoietin)
500 IU
RUO
EP; EPO; Epoetin; erythropoietin; MGC138142; MVCD2
EPO
Protein
Human
APPRLICDSR VLERYLLEAK EAENITTGCA EHCSLNENIT VPDTKVNFYA WKRMEVGQQA VEVWQGLALL SEAVLRGQAL LVNSSQPWEP LQLHVDKAVS GLRSLTTLLR ALGAQKEAIS PPDAASAAPL RTITADTFRK LFRVYSNFLR GKLKLYTGEA CRTGDR
Carrier-Free
<0.1 ng/μg
CHO cells
Recombinant, Ligand
EPO
Gibco
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions