missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human EPO ELISA Kit
ELISA
$882.00 - $6220.00
Specifications
| Accession Number | EP; EPO; epoetin; Erythropoietin; MGC138142; MVCD2 |
|---|---|
| Assay Range | 1.6 to 100 |
| Assay Sensitivity | 0.14 mIU/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
BMS20352
|
Invitrogen™
BMS20352 |
96 Tests |
Each for $882.00
|
|
|||||
|
BMS20352TEN
|
Invitrogen™
BMS20352TEN |
10 x 96 Tests |
Pack of 10 Each for $6,220.00
|
|
|||||
Description
The Human EPO ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative detection of human EPO.
Erythropoietin (EPO) protein is a growth factor essential for the survival, proliferation, and differentiation of erythroid precursors. EPO is not responsible for early commitment to the lineage, and instead induces the final maturation of partially differentiated erythroid cells and the production of red cell-specific proteins, such as hemoglobin. The EPO receptor (EPOR) is present in higher levels on precursor cells, and expression diminishes with cell maturation.Hepatocytes are the main source of EPO in the fetus, followed by interstitial cells of the kidney after birth and through adulthood. EPO expression is upregulated during hypoxia, and low levels of transcription have also been detected in lung and brain tissue under these conditions. Recent data suggest that EPO may also play a role in the protection of neuronal tissue from damage and initiation of repair.Specifications
| EP; EPO; epoetin; Erythropoietin; MGC138142; MVCD2 | |
| 0.14 mIU/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| EPO | |
| 6.2 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Epo | |
| 3 hrs 30 mins |
| 1.6 to 100 | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 2056 | |
| 4.3% | |
| Pre-coated 96 well plate, Standard (2 vials), Standard (2 vials), Assay Buffer concentrate, Streptavidin-HRP, Wash Buffer, Wash Buffer, Substrate Solution, Stop Solution, Adhesive Plate Covers, Control High | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |