missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EMP2 (aa 25-60) Control Fragment Recombinant Protein

Catalog No. RP91201
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91201 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91201 Supplier Invitrogen™ Supplier No. RP91201
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (61%), Rat (61%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53220 (PA5-53220. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Epithelial membrane protein-2 (EMP2) is a member of the four transmembrane superfamily (TM4SF) and is thought to mediate trafficking of diverse proteins such as alpha6beta1 integrin and MHC class I to lipid raft microdomains. EMP2 has also recently been recognized as a putative tumor suppressor gene in certain model systems.

Specifications

Accession Number P54851
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 2013
Name Human EMP2 (aa 25-60) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias EMP2; EMP-2; Epithelial membrane protein 2; MGC9056; Protein XMP; Unknown (protein for MGC:137044); XMP
Common Name EMP2
Gene Symbol EMP2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DNAWWVGDEFFADVWRICTNNTNCTVINDSFQEYST
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less