missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human eIF3l (aa 400-537) Control Fragment Recombinant Protein

Catalog No. RP91347
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91347 Supplier Invitrogen™ Supplier No. RP91347

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51799 (PA5-51799. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis (PubMed:17581632, PubMed:25849773, PubMed:27462815). The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S pre-initiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post-termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation (PubMed:17581632). The eIF-3 complex specifically targets and initiates translation of a subset of mRNAs involved in cell proliferation, including cell cycling, differentiation and apoptosis, and uses different modes of RNA stem-loop binding to exert either translational activation or repression (PubMed:25849773). [UniProt]

Specifications

Accession Number Q9Y262
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51386
Name Human eIF3l (aa 400-537) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 0610011H21Rik; 66 kDa tyrosine-rich heat shock protein; 67 kDa polymerase-associated factor; D15N1e; eIEF associated protein HSPC021; EIF3EIP; Eif3ip; EIF3L; EIF3S11; Eif3s6ip; Eukaryotic translation initiation factor 3 subunit 6-interacting protein; eukaryotic translation initiation factor 3 subunit E-interacting protein; Eukaryotic translation initiation factor 3 subunit L; eukaryotic translation initiation factor 3, subunit 6 interacting protein; eukaryotic translation initiation factor 3, subunit L; HSP-66 Y; HSPC021; HSPC025; MSTP005; Paf67
Common Name eIF3l
Gene Symbol EIF3L
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RMQKGDPQVYEELFSYSCPKFLSPVVPNYDNVHPNYHKEPFLQQLKVFSDEVQQQAQLSTIRSFLKLYTTMPVAKLAGFLDLTEQEFRIQLLVFKHKMKNLVWTSGISALDGEFQSASEVDFYIDKDMIHIADTKVAR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less