missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EIF1AD (aa 75-145) Control Fragment Recombinant Protein

Catalog No. RP97490
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP97490 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP97490 Supplier Invitrogen™ Supplier No. RP97490

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63608 (PA5-63608. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications

Accession Number Q8N9N8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84285
Name Human EIF1AD (aa 75-145) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2010003J03Rik; AI131644; Eif1ad; eukaryotic translation initiation factor 1 A domain containing; eukaryotic translation initiation factor 1 A domain-containing protein; Haponin; probable RNA-binding protein EIF1AD; RGD1304686
Common Name EIF1AD
Gene Symbol EIF1AD
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EEGEKVKAEISFVLCKDHVRSLQKEGFWPEAFSEVAEKHNNRNRQTQPELPAEPQLSGEESSSEDDSDLFV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less