missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human EFHD2 (aa 3-31) Control Fragment Recombinant Protein

Catalog No. RP98470
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98470 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98470 Supplier Invitrogen™ Supplier No. RP98470
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61846 (PA5-61846. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

EFHD2, also known as Swiprosin-1 or SWS1, is an EF-hand and coiled-coil-containing adaptor protein that plays a role in lymphocyte physiology. EFHD2 exhibits the highest expression in CD8+ T cells and immature B cells. It provides a membrane scaffold that is required for the Syk-, SLP-65-, and PLC-gamma2-dependent B-cell receptor (BCR)-induced calcium flux. EFHD2 may also regulate BCR-induced immature and primary B-cell apoptosis. It controls spontaneous apoptosis through the regulation of BCL2L1 abundance. Also, EFHD2 plays a role as negative regulator of the canonical NF-kappa-B-activating branch.

Specifications

Accession Number Q96C19
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79180
Name Human EFHD2 (aa 3-31) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2600015J22Rik; AA408606; D4Wsu27e; EF hand domain containing 2; EF hand domain family, member D2; EF-hand domain family member D2; EF-hand domain family, member D2; EF-hand domain-containing protein D2; Efhd2; MGC4342; RP3-467K16.3; swiprosin 1; swiprosin-1; Sws1; testicular tissue protein Li 62
Common Name EFHD2
Gene Symbol Efhd2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TDELATKLSRRLQMEGEGGGETPEQPGLN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less