missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human EEF1A2 (aa 359-390) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP100016
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84172 (PA5-84172. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes an isoform of the alpha subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This isoform (alpha 2) is expressed in brain, heart and skeletal muscle, and the other isoform (alpha 1) is expressed in brain, placenta, lung, liver, kidney, and pancreas. This gene may be critical in the development of ovarian cancer. [provided by RefSeq, Mar 2014]Specifications
| Q05639 | |
| Blocking Assay, Control | |
| 1917 | |
| 100 μL | |
| Eef1a; EEF1A2; eEF1A-2; EEF1AL; EF1A; EF-1-alpha-2; EIEE33; elongation factor 1 A-2; elongation factor 1-alpha 2; Eukaryotic elongation factor 1 A-2; eukaryotic translation elongation factor 1 alpha 2; HS1; MRD38; Ps10; RATPS10; S1; statin S1; Statin-like protein; statin-S1; STN; Stnl; wasted; wst | |
| EEF1A2 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human EEF1A2 (aa 359-390) Control Fragment | |
| RUO | |
| EEF1A2 | |
| Unconjugated | |
| Recombinant | |
| PVIDCHTAHIACKFAELKEKIDRRSGKKLEDN | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |