missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DYRK1A (aa 2-69) Control Fragment Recombinant Protein

Catalog No. RP91461
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91461 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91461 Supplier Invitrogen™ Supplier No. RP91461
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-110809 (PA5-110809. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

DYRK1A is a dual specificity kinase involved in cell proliferation. The gene for DYRK1A is located in the Down syndrome critical region of chromosome 21, and has been implicated in the pathogenesis of this disease. DYRK1A catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.

Specifications

Accession Number Q13627
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1859
Name Human DYRK1A (aa 2-69) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2310043O08Rik; D16Ertd272e; D16Ertd493e; dual specificity tyrosine phosphorylation regulated kinase 1 A; dual specificity tyrosine-(Y)-phosphorylation regulated kinase 1 A; dual specificity tyrosine-phosphorylation-regulated kinase 1 A; Dual specificity YAK1-related kinase; dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1 A; Dyrk; DYRK1; Dyrk1a; ENSMUSG00000074897; Gm10783; hMNB; HP86; mmb; MNB; mnb protein kinase homolog hp86; MNB/DYRK protein kinase; MNBH; MP86; MRD7; OTTHUMP00000109090; Protein kinase minibrain homolog; PSK47; RP86; serine/threonine kinase MNB; serine/threonine-specific protein kinase
Common Name DYRK1A
Gene Symbol DYRK1A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HTGGETSACKPSSVRLAPSFSFHAAGLQMAGQMPHSHQYSDRRQPNISDQQVSALSYSDQIQQPLTNQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less