missing translation for 'onlineSavingsMsg'
Learn More

ProSci Human Duodenum Lysate

Catalog No. 16916820
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16916820 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16916820 Supplier ProSci Supplier No. 13640100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Total protein normal duodenum - Human

  • Duodenum tissue lysate was prepared by homogenization in lysis buffer (10 mM HEPES pH7.9, 1.5 mM MgCl2, 10 mM KCl, 1 mM ethylenediaminetetraacetic acid, 10% glycerol, 1% NP-40, and a cocktail of protease inhibitors)
  • Tissue and cell debris was removed by centrifugation
  • The product was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6.8, 12.5% glycerol, 1% sodium dodecylsulfate, 0.01% bromophenol blue) containing 50 mM DTT.

Specifications

Tissue Duodenum
Quantity 0.1 mg
Tissue Condition Normal
Host Species Human
Concentration 2 mg/mL
Protein State Denatured