missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DSCC1 (aa 298-391) Control Fragment Recombinant Protein

Recombinant Protein

Supplier:  Invitrogen™ RP93796

Catalog No. RP93796


Only null left
Explore available promotions

Description

Description

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54885 (PA5-54885. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CHTF18 (MIM 613201), CHTF8 (MIM 613202), and DSCC1 arecomponents of an alternative replication factor C (RFC) (see MIM600404) complex that loads PCNA (MIM 176740) onto DNA during Sphase of the cell cycle.
Specifications

Specifications

Q9BVC3
Blocking Assay, Control
79075
100 μL
2010006I05Rik; 2600005O03Rik; DCC1; defective in sister chromatid cohesion 1 homolog; defective in sister chromatid cohesion 1 homolog (S. cerevisiae); defective in sister chromatid cohesion protein 1 homolog; DNA replication and sister chromatid cohesion 1; DSCC1; RGD1561749; Sister chromatid cohesion protein DCC1; UNQ9337/PRO34008
DSCC1
Human
His-ABP-tag
-20°C, Avoid Freeze/Thaw Cycles
Liquid
≥5.0 mg/mL
1 M urea, PBS with no preservative; pH 7.4
Human DSCC1 (aa 298-391) Control Fragment
RUO
DSCC1
Unconjugated
Recombinant
EGMVTSLDQLKGLALVDRHSRPEIIFLLKVDDLPEDNQERFNSLFSLREKWTEEDIAPYIQDLCGEKQTIGALLTKYSHSSMQNGVKVYNSRRP
E. coli
>80% by SDS-PAGE and Coomassie blue staining
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions