missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Accession Number | 4921507G06Rik; 4930435C18Rik; BB239540; Down syndrome cell adhesion molecule 2; Down syndrome cell adhesion molecule like 1; Down syndrome cell adhesion molecule-like 1; Down syndrome cell adhesion molecule-like protein 1; Down syndrome cell adhesion molecule-like protein 1 homolog; Downs syndrome cell adhesion molecule like 1; DS cell adhesion molecule like 1; DSCAM2; DSCAML1; DSCAM-like 1; KIAA1132; mKIAA1132 |
|---|---|
| Assay Range | 0.82 to 200 ng/mL |
| Assay Sensitivity | 0.82 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
EH164RBX5
|
Invitrogen™
EH164RBX5 |
5 x 96 Tests |
Each for $4,330.00
|
|
|||||
EH164RB
|
Invitrogen™
EH164RB |
96 Tests |
Each for $872.00
|
|
|||||
Description
Human DSCAM-L1/DSCAM2 quantitates human DSCAM-L1/DSCAM2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human DSCAM-L1/DSCAM2.
DSCAML1 encodes a protein that is a member of the Ig superfamily of cell adhesion molecules and is involved in neuronal differentiation. The encoded membrane-bound protein localizes to the cell surface, where it forms aggregates that repel neuronal processes of the same cell type.Specifications
| 4921507G06Rik; 4930435C18Rik; BB239540; Down syndrome cell adhesion molecule 2; Down syndrome cell adhesion molecule like 1; Down syndrome cell adhesion molecule-like 1; Down syndrome cell adhesion molecule-like protein 1; Down syndrome cell adhesion molecule-like protein 1 homolog; Downs syndrome cell adhesion molecule like 1; DS cell adhesion molecule like 1; DSCAM2; DSCAML1; DSCAM-like 1; KIAA1132; mKIAA1132 | |
| 0.82 ng/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| DSCAML1 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| DSCAML1 | |
| 4 hrs 45 mins |
| 0.82 to 200 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 57453 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |