missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DSC-3 ELISA Kit

Catalog No. p-7219343
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH162RBX5 5 x 96 Tests
EH162RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH162RBX5 Supplier Invitrogen™ Supplier No. EH162RBX5
Only null left

ELISA

Human DSC-3 quantitates human DSC-3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human DSC-3.

Desmosomes are cell-cell junctions between epithelial, myocardial, and certain other cell types. Currently, three desmoglein subfamily members have been identified and all are members of the cadherin cell adhesion molecule superfamily. These desmoglein gene family members are located in a cluster on chromosome 18. This protein has been identified as the autoantigen of the autoimmune skin blistering disease pemphigus vulgaris.

Specifications

Accession Number 5430426I24Rik; ARVC10; ARVD10; Cadherin family member 3; CDHF3; CDHF5; CDHF6; CMD1BB; desmocollin 3; desmocollin-3; Desmocollin-4; desmoglein-2; desmoglein-3; DSC; DSC1; DSC2; DSC3; DSC4; HDGC; HT-CP; PVA
Assay Range 0.16 to 40 ng/mL
Assay Sensitivity 0.16 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 1825
Gene Symbol Dsc3
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target Desmocollin 3
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less