missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DREV (aa 236-315) Control Fragment Recombinant Protein

Catalog No. RP98801
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98801 Supplier Invitrogen™ Supplier No. RP98801

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62814 (PA5-62814. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications

Accession Number Q9H1A3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 51108
Name Human DREV (aa 236-315) Control Fragment
Quantity 100 μL
Gene Alias 0610012D09Rik; AA517660; CGI-81; CTB-31N19.3; DORA reverse strand protein; DORA reverse strand protein 1; Drev; DREV1; methyltransferase like 9; methyltransferase-like protein 9; Mettl9; MNCb-5680; p53 activated protein 1; PAP1
Common Name DREV
Gene Symbol Mettl9
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VILALVLPFHPYVENVGGKWEKPSEILEIKGQNWEEQVNSLPEVFRKAGFVIEAFTRLPYLCEGDMYNDYYVLDDAVFVL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less