missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DOCK1 (aa 1614-1693) Control Fragment Recombinant Protein

Catalog No. RP106449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP106449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP106449 Supplier Invitrogen™ Supplier No. RP106449

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84144 (PA5-84144. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a loosely associated peripheral membrane protein related to the LanC family of bacterial membrane-associated proteins involved in the biosynthesis of antimicrobial peptides. This protein may play a role as a peptide-modifying enzyme component in eukaryotic cells. Previously considered a member of the G-protein-coupled receptor superfamily, this protein is now in the LanC family. Multiple alternatively spliced variants, encoding the same protein, have been identified.

Specifications

Accession Number Q14185
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1793
Name Human DOCK1 (aa 1614-1693) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 180 kDa protein downstream of CRK; 9130006G06Rik; AI854900; ced5; D630004B07Rik; dedicator of cytokinesis 1; dedicator of cyto-kinesis 1; dedicator of cytokinesis protein 1; dock 180; DOCK1; DOCK180; DOwnstream of CrK; RGD1566072; RP11-82L9.1
Common Name DOCK1
Gene Symbol DOCK1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VRIMPSSLDDRRGSRPRSMVRSFTMPSSSRPLSVASVSSLSSDSTPSRPGSDGFALEPLLPKKMHSRSQDKLDKDDLEKE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less