missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DNMBP (aa 389-468) Control Fragment Recombinant Protein

Catalog No. RP106830
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP106830 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP106830 Supplier Invitrogen™ Supplier No. RP106830

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (63%), Rat (63%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66148 (PA5-66148. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

DNMBP belongs to the DBL.

Specifications

Accession Number Q6XZF7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23268
Name Human DNMBP (aa 389-468) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2410003L07Rik; 2410003M15Rik; ARHGEF36; Dnmbp; dynamin binding protein; dynamin-binding protein; Kiaa1010; Scaffold protein Tuba; TUBA
Common Name DNMBP
Gene Symbol DNMBP
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GVEWEMPLATDSPTSDPTEVVNGISSQPQVPFHPNLQKSQYYSTVGGSHPHSEQYPDLLPLEARTRDYASLPPKRMYSQL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less