missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DNAJC3 (aa 226-307) Control Fragment Recombinant Protein

Catalog No. RP97911
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP97911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP97911 Supplier Invitrogen™ Supplier No. RP97911

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83458. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein with multiple tetratricopeptide repeat (TPR) motifs as well as the highly conserved J domain found in DNAJ chaperone family members. It is a member of the tetratricopeptide repeat family of proteins and acts as an inhibitor of the interferon-induced, dsRNA-activated protein kinase (PKR).

Specifications

Accession Number Q13217
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 5611
Name Human DNAJC3 (aa 226-307) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AA408985; ACPHD; AU067833; DnaJ (Hsp40) homolog, subfamily C, member 3; DnaJ (Hsp40) homolog, subfamily C, member 3A; DnaJ (Hsp40) homolog, subfamily C, member 3B; DnaJ heat shock protein family (Hsp40) member C3; dnaJ homolog subfamily C member 3; DNAJC3; Dnajc3a; Dnajc3b; Endoplasmic reticulum DNA J domain-containing protein 6; ERdj6; ER-resident protein ERdj6; FLJ21288; HP58; Interferon-induced, double-stranded RNA-activated protein kinase inhibitor; mp58; p58; p58IPK; PRKRI; Protein kinase inhibitor of 58 kDa; protein kinase inhibitor p58; protein kinase, interferon inducible double stranded RNA dependent inhibitor; protein-kinase, interferon-inducible double stranded RNA dependent inhibitor
Common Name DNAJC3
Gene Symbol DNAJC3
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence YKISTLYYQLGDHELSLSEVRECLKLDQDHKRCFAHYKQVKKLNKLIESAEELIRDGRYTDATSKYESVMKTEPSIAEYTVR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less