missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DLEC/CLEC4C/BDCA-2 ELISA Kit

Catalog No. EH156RBX5
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH156RBX5 5 x 96 Tests
EH156RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH156RBX5 Supplier Invitrogen™ Supplier No. EH156RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions

ELISA

Human DLEC/CLEC4C/BDCA-2 quantitates human DLEC/CLEC4C/BDCA-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human DLEC/CLEC4C/BDCA-2.

This gene encodes a member of the C-type lectin/C-type lectin-like domain superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in dendritic cell function. Two transcript variants encoding distinct isoforms have been identified for this gene.

Specifications

Accession Number Q8WTT0
Assay Range 0.082-20 ng/mL
Assay Sensitivity 0.082 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias BDCA2; BDCA-2; Blood dendritic cell antigen 2; blood dendritic cell antigen 2 protein; CD303; CD303 antigen; CLEC4C; CLECSF11; CLECSF7; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 11; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 7; C-type lectin domain family 4 member C; C-type lectin domain family 4, member C; C-type lectin domain family 4, member C, isoform 1; C-type lectin superfamily member 7; dendritic cell lectin b; dendritic lectin; DLEC; HECL; PRO34150; UNQ9361/PRO34150BDCA2; BDCA-2; Blood dendritic cell antigen 2; blood dendritic cell antigen 2 protein; CD303; CD303 antigen; CLEC4C; CLECSF11; CLECSF7; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 11; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 7; C-type lectin domain family 4 member C; C-type lectin domain family 4, member C; C-type lectin domain family 4, member C, isoform 1; C-type lectin superfamily member 7; dendritic cell lectin b; dendritic lectin; DLEC; HECL; PRO34150; UNQ9361/PRO34150;
Gene ID (Entrez) 170482
Gene Symbol CLEC4C
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target CLEC4C
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less