missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DIS3L (aa 103-201) Control Fragment Recombinant Protein

Catalog No. RP98155
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98155 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98155 Supplier Invitrogen™ Supplier No. RP98155

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (91%), Rat (91%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59739 (PA5-59739. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

DIS3L is a probable exonuclease. It has four isoforms.

Specifications

Accession Number Q8TF46
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 115752
Name Human DIS3L (aa 103-201) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AV340375; DIS3 like exosome 3'-5' exoribonuclease; DIS3 mitotic control homolog (S. cerevisiae)-like; DIS3 mitotic control homolog-like; Dis3l; DIS3L1; DIS3-like exonuclease 1; DIS3-like exosome 3'-5' exoribonuclease; KIAA1955; RGD1308959
Common Name DIS3L
Gene Symbol DIS3L
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LRNLLKDARHDCILFANEFQQCCYLPRERGESMEKWQTRSIYNAAVWYYHHCQDRMPIVMVTEDEEAIQQYGSETEGVFVITFKNYLDNFWPDLKAAHE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less