missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DDX19A (aa 15-39) Control Fragment Recombinant Protein

Catalog No. RP100363
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP100363 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP100363 Supplier Invitrogen™ Supplier No. RP100363

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60962 (PA5-60962. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ATP-dependent RNA helicase involved in mRNA export from the nucleus.

Specifications

Accession Number Q9NUU7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55308
Name Human DDX19A (aa 15-39) Control Fragment
Quantity 100 μL
Gene Alias ATP-dependent RNA helicase DD x 19 A; DBP5; Dd x 19; DD x 19 A; DD x 19-DD x 19 L; DD x 19 L; DD x 19-like protein; DEAD (Asp-Glu-Ala-As) box polypeptide 19 A; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19; DEAD (Asp-Glu-Ala-Asp) box polypeptide 19 A; DEAD box protein 19 A; DEAD box RNA helicase DEAD5; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 19; DEAD-box helicase 19 A; Eif4a-rs1; eukaryotic translation initiation factor 4 A related sequence 1; eukaryotic translation initiation factor 4 A-related sequence 1; mDEAD5; RNA helicase; ZD10B; zinc responsive protein ZD10B
Common Name DDX19A
Gene Symbol DDX19A
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AAVKSMTNLQIKEEKVKADTNGIIK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less