missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human DDA1 (aa 82-102) Control Fragment Recombinant Protein

Catalog No. RP106616
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP106616 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP106616 Supplier Invitrogen™ Supplier No. RP106616
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65934 (PA5-65934. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Functions as a component of numerous distinct DCX (DDB1-CUL4-X-box) E3 ubiquitin-protein ligase complexes which mediate the ubiquitination and subsequent proteasomal degradation of target proteins (PubMed:17452440, PubMed:28437394, PubMed:28302793, PubMed:31686031, PubMed:31819272). In the DCX complexes, acts as a scaffolding subunit required to stabilize the complex (PubMed:31686031, PubMed:31819272). [UniProt]

Specifications

Accession Number Q9BW61
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 79016
Name Human DDA1 (aa 82-102) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1500034J01Rik; 1700095J19Rik; C19orf58; cross-immune reaction antigen PCIA1; Dda1; DET1 and DDB1 associated 1; DET1- and DDB1-associated protein 1; PCIA1; PCIA-1; Placenta cross-immune reaction antigen 1; R74921
Common Name DDA1
Gene Symbol DDA1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ESSAPPRKVARTDSPDMHEDT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less