missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Cytokeratin 23 Control Fragment Recombinant Protein

Catalog No. RP89653
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP89653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP89653 Supplier Invitrogen™ Supplier No. RP89653
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53524 (PA5-53524. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.

Specifications

Accession Number Q9C075
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 25984
Name Human Cytokeratin 23 Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C HAIK1; CK23; CK-23; cytokeratin 23; cytokeratin-23; DKFZP434G032; HAIK1; histone deacetylase inducible keratin 23; hyperacetylation-inducible type I keratin; K23; Ka23; keratin 23; keratin 23 (histone deacetylase inducible); keratin 23, type I; keratin complex 1, acidic, gene 23; keratin, type I cytoskeletal 23; Keratin-23; Krt1-23; KRT23; MGC26158; type I intermediate filament cytokeratin; type I keratin KA23
Common Name Cytokeratin 23
Gene Symbol KRT23
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KHHVPSDFNVNVKVDTGPREDLIKVLEDMRQEYELIIKKKHRDLDTWYKEQSAAMSQEAASPATVQSRQGDIHELKRTFQALEIDLQAQYSTKSALENMLSETQSRYSCKLQDMQEIISHYEEELTQLR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less