missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Cystatin A ELISA Kit
ELISA
Supplier: Invitrogen™ EH140RB
Description
Human Cystatin A quantitates human Cystatin A in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cystatin A.
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins, and kininogens. This gene encodes a stefin that functions as a cysteine protease inhibitor, forming tight complexes with papain and the cathepsins B, H, and L. The protein is one of the precursor proteins of cornified cell envelope in keratinocytes and plays a role in epidermal development and maintenance. Stefins have been proposed as prognostic and diagnostic tools for cancer.Chemical Identifiers
| AREI; Csta; Csta1; cystatin A; cystatin A (stefin A); cystatin A1; cystatin AS; cystatin-A; Cystatin-A, N-terminally processed; Cystatin-A1; Cystatin-AS; PSS4; Stefin A; Stefin-A; STF1; STFA | |
| 6.5 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| CSTA | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Cystatin A | |
| 4 hrs 45 mins |
| 6.5 to 1,500 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 1475 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
Specifications
| AREI; Csta; Csta1; cystatin A; cystatin A (stefin A); cystatin A1; cystatin AS; cystatin-A; Cystatin-A, N-terminally processed; Cystatin-A1; Cystatin-AS; PSS4; Stefin A; Stefin-A; STF1; STFA | |
| 6.5 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| CSTA | |
| <10 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |
| 6.5 to 1,500 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 1475 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| Cystatin A | |
| 4 hrs 45 mins |