missing translation for 'onlineSavingsMsg'
Learn More

Human CXCL4/PF4 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Catalog No. IC7952U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC7952U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC7952U100 Supplier Bio-Techne Supplier No. IC7952U100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

CXCL4/PF4 Monoclonal specifically detects CXCL4/PF4 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen CXCL4/PF4
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 170138
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P02776.2
Gene Alias chemokine (C-X-C motif) ligand 4, C-X-C motif chemokine 4, CXCL4iroplact, Iroplact, MGC138298, Oncostatin-A, PF4, PF-4, platelet factor 4, SCYB4oncostatin-A
Gene Symbols PF4
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL4/PF4 Glu32-Ser101 Accession # P02776.2
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5196
Test Specificity Detects human CXCL4/PF4 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) CXCL6, recombinant human (rh) CXCL6, rhCXCL7, rmCXCL13 or rhCCL21 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Product Type Antibody
Isotype IgG2b
Product Line R&D Systems
Show More Show Less

For Research Use Only (RUO)