missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CXCL2 (GRO beta) Recombinant Protein
Recombinant Protein
Supplier: Gibco™ PHC1074
Description
Human CXCL2 (GRO beta) Recombinant Protein for Bioactivity
Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.Specifications
| P19875 | |
| PBS with no preservative; pH 7.4 | |
| 19.3 kDa | |
| 7.4 | |
| 10 μg | |
| RUO | |
| C Cmotif chemokine; C X C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 2; CINC-2a; Cinc3; CINC-3; CXC; CXC motif chemokine; C-X-C motif chemokine 2; C-X-C motif chemokine ligand 2; CXCL; CXCL2; Cytokine-induced neutrophil chemoattractant 3; GRO b; gro beta; GRO β; GRO2; GRO2 oncogene; GROb; GRObeta; gro-beta; GRO-beta(5-73); GRO-beta-T; Growth-regulated protein beta; GROβ; Hematopoietic synergistic factor; HSF; Macrophage inflammatory protein 2; macrophage inflammatory protein 2-alpha; melanoma growth stimulatory activity beta; MGSA beta; Mgsa-b; MIP 2 a; MIP 2 alpha; MIP 2 α; Mip2; MIP-2; MIP2 a; MIP2 α; MIP2A; MIP-2a; MIP2-alpha; SB-251353; Scyb; Scyb2; small inducible cytokine subfamily, member 2 | |
| Cxcl2 | |
| Protein | |
| Human | |
| MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEI GVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS. | |
| ED50 < 3 ng/mL; determined by the dose-dependent stimulation BaF3 indicator cells | |
| Approved for shipment on Wet or Dry Ice | |
| Chemokines & Receptors | |
| Lyophilized | |
| Inflammation, Oncology | |
| >96% by SDS-PAGE |
| Bioactivity | |
| 2920 | |
| Human CXCL2 (GRO beta) | |
| Purified | |
| -20°C | |
| <0.1 ng/μg | |
| CXCL2 | |
| ED50 < 3 ng/mL; determined by the dose-dependent stimulation BaF3 indicator cells | |
| Unconjugated | |
| Recombinant | |
| -20°C | |
| <0.1 ng/μg | |
| E. coli | |
| Recombinant, Ligand | |
| GROâKCâCINC Chemokines | |
| Gibco |