missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CSN3 (CASK) (aa 84-145) Control Fragment Recombinant Protein

Catalog No. RP96738
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP96738 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP96738 Supplier Invitrogen™ Supplier No. RP96738

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (34%), Rat (34%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57480 (PA5-57480. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Kappa-casein (CSN3) stabilizes micelle formation, preventing casein precipitation in milk.

Specifications

Accession Number P07498
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1448
Name Human CSN3 (CASK) (aa 84-145) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias casein kappa; CASK; CNS10; CSN10; CSN3; CSNK; kappa-casein; KCA
Common Name CSN3 (CASK)
Gene Symbol CSN3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PAVVRPHAQIPQRQYLPNSHPPTVVRRPNLHPSFIAIPPKKIQDKIIIPTINTIATVEPTPA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less