missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CRHSP-24 (aa 1-31) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP101377
Description
Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62515 (PA5-62515. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
CRHSP-24 is a protein coding gene.Specifications
| Q9Y2V2 | |
| Blocking Assay, Control | |
| 23589 | |
| 100 μL | |
| 1200011K09Rik; calcineurin substrate CRHSP-24; calcium regulated heat stable protein 1; calcium regulated heat stable protein 1, 24 kDa; calcium-regulated heat stable protein 1; calcium-regulated heat-stable protein (24 kD); calcium-regulated heat-stable protein 1; Calcium-regulated heat-stable protein of 24 kDa; Carhsp1; CRHSP24; Crhsp-24; CSDC1; D16Ertd465e | |
| CARHSP1 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human CRHSP-24 (aa 1-31) Control Fragment | |
| RUO | |
| CRHSP-24 | |
| Unconjugated | |
| Recombinant | |
| MSSEPPPPPQPPTHQASVGLLDTPRSRERSP | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |