missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CPSF4L (aa 7-61) Control Fragment Recombinant Protein

Catalog No. RP99971
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99971 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99971 Supplier Invitrogen™ Supplier No. RP99971

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (80%), Rat (80%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60603 (PA5-60603. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications

Accession Number A6NMK7
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 642843
Name Human CPSF4L (aa 7-61) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 0610010C04Rik; 1500000C01Rik; cleavage and polyadenylation specific factor 4 like; cleavage and polyadenylation specific factor 4-like; Cpsf4l; D11Ertd636e; putative cleavage and polyadenylation specificity factor subunit 4-like protein; RGD1306397
Common Name CPSF4L
Gene Symbol Cpsf4l
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GLERFTFAFEKDVEMQKGTGLLPFQGMDKSASAVCNFFTKGLCEKGKLCPFRHDR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less