missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Contactin 3 (aa 373-502) Control Fragment Recombinant Protein

Catalog No. RP102168
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP102168 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP102168 Supplier Invitrogen™ Supplier No. RP102168

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51900. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Contactin 3 is a type of contactin that mediate cell surface interactions during nervous system development. Has some neurite outgrowth-promoting activity.

Specifications

Accession Number Q9P232
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 5067
Name Human Contactin 3 (aa 373-502) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias BIG-1; Brain-derived immunoglobulin superfamily protein 1; Cntn3; contactin 3; contactin 3 (plasmacytoma associated); contactin-3; KIAA1496; Pang; PCS; Plasmacytoma-associated neuronal glycoprotein; Plasmacytoma-associated neuronal glycoprotein (neural cell adhesion molecule BIG-1)
Common Name Contactin 3
Gene Symbol CNTN3
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ISNLSVTDSGMFQCIAENKHGLVYSSAELKVVASAPDFSKNPMKKLVQVQVGSLVSLDCKPRASPRALSSWKKGDVSVQEHERISLLNDGGLKIANVTKADAGTYTCMAENQFGKANGTTHLVVTEPTRI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less