missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human COG6 (aa 325-425) Control Fragment Recombinant Protein

Catalog No. RP97946
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP97946 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP97946 Supplier Invitrogen™ Supplier No. RP97946

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59163. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a subunit of the conserved oligomeric Golgi complex that is required for maintaining normal structure and activity of the Golgi apparatus. The encoded protein is organized with conserved oligomeric Golgi complex components 5, 7 and 8 into a sub-complex referred to as lobe B. Alternative splicing results in multiple transcript variants.

Specifications

Accession Number Q9Y2V7
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 57511
Name Human COG6 (aa 325-425) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 4933405E16Rik; AU018618; CDG2L; COD2; COG complex subunit 6; COG6; complexed with Dor1p 2; component of oligomeric golgi complex 6; conserved oligomeric Golgi complex protein 6; conserved oligomeric Golgi complex subunit 6; Conserved oligomeric Golgi complex subunit 6-like protein; hypothetical protein MGC56117; KIAA1134; LOW QUALITY PROTEIN: conserved oligomeric Golgi complex subunit 6; mKIAA1134; SHNS; si:ch211-203f4.1; testicular tissue protein Li 41; zgc:56117
Common Name COG6
Gene Symbol COG6
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HLEALLKHVTTQGVEENIQEVVGHITEGVCRPLKVRIEQVIVAEPGAVLLYKISNLLKFYHHTISGIVGNSATALLTTIEEMHLLSKKIFFNSLSLHASKL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less