missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human COG3 (aa 607-705) Control Fragment Recombinant Protein

Catalog No. RP108403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP108403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP108403 Supplier Invitrogen™ Supplier No. RP108403

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84200 (PA5-84200. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a component of the conserved oligomeric Golgi (COG) complex which is composed of eight different subunits and is required for normal Golgi morphology and localization. Defects in the COG complex result in multiple deficiencies in protein glycosylation. The protein encoded by this gene is involved in ER-Golgi transport.

Specifications

Accession Number Q96JB2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 83548
Name Human COG3 (aa 607-705) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias COG complex subunit 3; COG3; component of oligomeric golgi complex 2; component of oligomeric golgi complex 3; Conserved oligomeric Golgi complex subunit 3; E430004N23; p94; SEC34; tethering factor SEC34; vesicle-docking protein SEC34 homolog
Common Name COG3
Gene Symbol COG3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DGQLFLIKHLLILREQIAPFHTEFTIKEISLDLKKTRDAAFKILNPMTVPRFFRLNSNNALIEFLLEGTPEIREHYLDSKKDVDRHLKSACEQFIQQQT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less