missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Coagulation Factor III ELISA Kit
ELISA
$890.00 - $4,420.00
Specifications
| Accession Number | AA409063; CD142; Cf3; Cf-3; Coagulation factor III; coagulation factor III (thromboplastin, tissue factor); coagulation factor III, tissue factor; F3; TF; TFA; Thromboplastin; Tissue factor |
|---|---|
| Assay Range | 6 to 1,500 pg/mL |
| Assay Sensitivity | 6 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
EH116RBX5
|
Invitrogen™
EH116RBX5 |
5 x 96 Tests |
Each for $4,420.00
|
|
|||||
EH116RB
|
Invitrogen™
EH116RB |
96 Tests |
Each for $890.00
|
|
|||||
Description
Human Coagulation Factor III quantitates human Coagulation Factor III in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Coagulation Factor III.
This gene encodes coagulation factor III which is a cell surface glycoprotein. This factor enables cells to initiate the blood coagulation cascades, and it functions as the high-affinity receptor for the coagulation factor VII. The resulting complex provides a catalytic event that is responsible for initiation of the coagulation protease cascades by specific limited proteolysis. Unlike the other cofactors of these protease cascades, which circulate as nonfunctional precursors, this factor is a potent initiator that is fully functional when expressed on cell surfaces. There are 3 distinct domains of this factor: extracellular, transmembrane, and cytoplasmic. This protein is the only one in the coagulation pathway for which a congenital deficiency has not been described.Specifications
| AA409063; CD142; Cf3; Cf-3; Coagulation factor III; coagulation factor III (thromboplastin, tissue factor); coagulation factor III, tissue factor; F3; TF; TFA; Thromboplastin; Tissue factor | |
| 6 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| F3 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| CD142 | |
| 4 hrs 45 mins |
| 6 to 1,500 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 2152 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |