missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CNTF, Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF45013500
Encompass
Change view
Click to view available options
Quantity:
100 μg
20 μg
500 μg
1 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF45013500 500 μg
AF45013100 100 μg
AF450131MG 1 mg
AF4501320UG 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AF45013500 Supplier Gibco™ Supplier No. AF45013500UG
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant Protein

Recombinant Human CNTF is synthesized as a 199 amino acid polypeptide (22.8 kDa) lacking a hydrophobic N-terminal signal for secretion. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a polypeptide hormone whose actions appear to be restricted to the nervous system where it promotes neurotransmitter synthesis and neurite outgrowth in certain neuronal populations. The protein is a potent survival factor for neurons and oligodendrocytes and may be relevant in reducing tissue destruction during inflammatory attacks. A mutation in this gene, which results in aberrant splicing, leads to ciliary neurotrophic factor deficiency, but this phenotype is not causally related to neurologic disease. A read-through transcript variant composed of ZFP91 and CNTF sequence has been identified, but it is thought to be non-coding. Read-through transcription of ZFP91 and CNTF has also been observed in mouse.

Specifications

Accession Number P26441
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 1270
Molecular Weight (g/mol) 22.8 kDa
Name Human CNTF, Animal-Free
Purification Method Purified
Quantity 500 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias AI429687; Ciliary neurotrophic factor; ciliary neurotropic factor; Cntf; HCNTF; H-CNTF
Common Name CNTF
Gene Symbol CNTF
Biological Activity Determined by its ability to stimulate proliferation of human TF-1 cells using an ED50 concentration range of 50-150 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence AFTEHSPLTP HRRDLCSRSI WLARKIRSDL TALTQSYVKH QGLNKNINLD SADGMPVAST DQWSQLTQAQ RLQQNLQAYR TFHVLLARLL QDQQVHFTPT QGDFHQAIHT LLLQVAAFAY QIQQLMILLQ YKIPRNQADG MPINVGDGGL FQKKLWGLKV LQQLSQWTVR SIHDLRFISS HQTGIPARGS HYIANNKKM
Content And Storage -20°C
Activity Determined by its ability to stimulate proliferation of human TF-1 cells using an ED50 concentration range of 50-150 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less