missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CNGA2 (aa 558-656) Control Fragment Recombinant Protein

Catalog No. RP91193
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91193 Supplier Invitrogen™ Supplier No. RP91193

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53274. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CNGA2 represents the alpha subunit of a cyclic nucleotide-gated olfactory channel. The encoded protein contains a carboxy-terminal leucine zipper that mediates channel formation.

Specifications

Accession Number Q16280
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 1260
Name Human CNGA2 (aa 558-656) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Cnca; CNCA1; CNCG2; Cncg4; cng channel; CNG channel 2; CNG channel alpha-2; CNG2; CNG-2; Cnga2; cyclic nucleotide gated channel 4; cyclic nucleotide gated channel alpha 2; cyclic nucleotide-gated cation channel 2; Cyclic nucleotide-gated channel alpha-2; cyclic nucleotide-gated olfactory channel; Cyclic nucleotide-gated olfactory channel subunit OCNC1; cyclic-nucleotide-gated cation channel 2; OCNC1; OCNCa; OCNCALPHA; OLFCH
Common Name CNGA2
Gene Symbol CNGA2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EAVTEYPDAKKVLEERGREILMKEGLLDENEVATSMEVDVQEKLGQLETNMETLYTRFGRLLAEYTGAQQKLKQRITVLETKMKQNNEDDYLSDGMNSP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less