missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CISD1 (aa 49-72) Control Fragment Recombinant Protein

Catalog No. RP107992
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP107992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP107992 Supplier Invitrogen™ Supplier No. RP107992

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67342. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a protein with a CDGSH iron-sulfur domain and has been shown to bind a redox-active [2Fe-2S] cluster. The encoded protein has been localized to the outer membrane of mitochondria and is thought to play a role in regulation of oxidation. Genes encoding similar proteins are located on chromosomes 4 and 17, and a pseudogene of this gene is located on chromosome 2.

Specifications

Accession Number Q9NZ45
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 55847
Name Human CISD1 (aa 49-72) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AU043990; AW743335; C10orf70; CDGSH iron sulfur domain 1; CDGSH iron sulfur domain-containing protein 1; CDGSH iron sulfur domain-containing protein-like protein; CDGSH iron-sulfur domain-containing protein 1; Cisd1; D10Ertd214e; hypothetical protein LOC393577; hypothetical protein LOC531444; MDS029; MGC14684; MitoNEET; RGD1309529; si:dkey-18n13.4; ZCD1; zgc:63561; zinc finger CDGSH-type domain 1; zinc finger, CDGSH-type domain 1
Common Name CISD1
Gene Symbol cisd1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IQKDNPKIVHAFDMEDLGDKAVYC
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less