missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CEP78 (aa 279-369) Control Fragment Recombinant Protein

Catalog No. RP92996
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP92996 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP92996 Supplier Invitrogen™ Supplier No. RP92996

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61823 (PA5-61823. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a centrosomal protein that is both required for the regulation of centrosome-related events during the cell cycle, and required for ciliogenesis. The encoded protein has an N-terminal leucine-rich repeat (LRR) domain with six consecutive LRR repeats, and a C-terminal coiled-coil domain. It interacts with the N-terminal catalytic domain of polo-like kinase 4 (PLK4) and colocalizes with PLK4 to the distal end of the centriole. Naturally occurring mutations in this gene cause defects in primary cilia that result in retinal degeneration and sensorineural hearing loss which are associated with cone-rod degeneration disease as well as Usher syndrome. Low expression of this gene is associated with poor prognosis of colorectal cancer patients.

Specifications

Accession Number Q5JTW2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84131
Name Human CEP78 (aa 279-369) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 5730599I05Rik; C9orf81; centrosomal protein 78; centrosomal protein 78 kDa; centrosomal protein of 78 kDa; Cep78; D030027P05; IP63
Common Name CEP78
Gene Symbol CEP78
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ALETNTTLVVLDIRKNPLIDHSMMKAVIKKVLQNGRSAKSEYQWITSPSVKEPSKTAKQKRRTIILGSGHKGKATIRIGLATKKPVSSGRK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less