missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CEP68 (aa 584-702) Control Fragment Recombinant Protein

Catalog No. RP98198
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP98198 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP98198 Supplier Invitrogen™ Supplier No. RP98198

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (68%), Rat (68%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59198 (PA5-59198. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Centrosomal protein of 68 kDa is a protein that in humans is encoded by the CEP68 gene. It is mapped to chromosome 2. CEP68 is required for centrosome cohesion. It decorates fibres emanating from the proximal ends of centrioles. CEP68 and rootletin depend both on each other for centriole association, and both also require CEP250 for their function.

Specifications

Accession Number Q76N32
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23177
Name Human CEP68 (aa 584-702) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6030463E10Rik; AI481761; BC027174; centrosomal protein 68; centrosomal protein 68 kDa; centrosomal protein of 68 kDa; Cep68; Kiaa0582; RGD1309101
Common Name CEP68
Gene Symbol CEP68
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SGLLKTRPSLPARLDRWPFSDPDVEGQLPRKGGEQGKESLVQCVKTFCCQLEELICWLYNVADVTDHGTAARSNLTSLKSSLQLYRQFKKDIDEHQSLTESVLQKGEILLQCLLENTPV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less