missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CENPT (aa 60-166) Control Fragment Recombinant Protein

Catalog No. RP104730
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP104730 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP104730 Supplier Invitrogen™ Supplier No. RP104730
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (73%), Rat (73%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-83540 (PA5-83540. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Component of the CENPA-NAC (nucleosome-associated) complex, a complex that plays a central role in assembly of kinetochore proteins, mitotic progression and chromosome segregation. The CENPA-NAC complex recruits the CENPA-CAD (nucleosome distal) complex and may be involved in incorporation of newly synthesized CENPA into centromeres. Part of a nucleosome-associated complex that binds specifically to histone H3-containing nucleosomes at the centromere, as opposed to nucleosomes containing CENPA. Component of the heterotetrameric CENP-T-W-S-X complex that binds and supercoils DNA, and plays an important role in kinetochore assembly. CENPT has a fundamental role in kinetochore assembly and function. It is one of the inner kinetochore proteins, with most further proteins binding downstream. Required for normal chromosome organization and normal progress through mitosis.

Specifications

Accession Number Q96BT3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 80152
Name Human CENPT (aa 60-166) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias C16orf56; CENPT; CENP-T; Centromere protein T; G630055P03Rik; ICEN22; Interphase centromere complex protein 22; RGD1309383
Common Name CENPT
Gene Symbol CENPT
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence RGRSHGARSVGRSAHIQASGHLEEQTPRTLLKNILLTAPESSILMPESVVKPVPAPQAVQPSRQESSCGSLELQLPELEPPTTLAPGLLAPGRRKQRLRLSVFQQGV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less