missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD63 Alexa Fluor 750-conjugated Antibody, R&D Systems™
Description
CD63 Monoclonal specifically detects CD63 in Human samples. It is validated for Flow Cytometry∣Flow Cytometry (Intracellular Staining).
Specifications
Specifications
| Antigen | CD63 |
| Applications | Flow Cytometry, Flow Cytometry (Intracellular Staining) |
| Classification | Monoclonal |
| Clone | 460305 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 750 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | P08962 |
| Gene Alias | CD63, CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, Lamp-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, Tspan30, TSPAN30granulophysin |
| Show More |
For Research Use Only (RUO)