missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TRAIL (TNFSF10) Recombinant Protein

Catalog No. 509293
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
509293 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 509293 Supplier Invitrogen™ Supplier No. BMS356
Only null left

Recombinant Protein

Description: Human TRAIL (TNF-Related Apoptosis Inducing Ligand), also called Apo2 Ligand, is a cytotoxic protein which activates rapid apoptosis in tumor cells, but not in normal cells. Human TRAIL/Apo2 Ligand is a 19.6 kDa protein, comprising the full-length of the TNF-like extracellular domain of TRAIL. Purity: >97%. Molecular Weight: 19.6 kDa.

TRAIL is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. TRAIL preferentially induces apoptosis in transformed and tumor cells, but does not appear to kill normal cells although it is expressed at a significant level in most normal tissues. TRAIL binds to several members of TNF receptor superfamily including TNFRSF10A/TRAILR1, TNFRSF10B/TRAILR2, TNFRSF10C/TRAILR3, TNFRSF10D/TRAILR4, and possibly also to TNFRSF11B/OPG. The activity of this protein may be modulated by binding to the decoy receptors TNFRSF10C/TRAILR3, TNFRSF10D/TRAILR4, and TNFRSF11B/OPG that cannot induce apoptosis. The binding of TRAIL protein to its receptors has been shown to trigger the activation of MAPK8/JNK, caspase 8, and caspase 3. Receptors for TRAIL include two death domain containing receptors, death receptor 4 (DR4) and DR5, as well as two decoy receptors, DcR1 and DcR2, lacking the intracellular signaling death domain. TRAIL is a type II membrane protein and expressed in a variety of human tissues. Like TNF and Fas ligand, TRAIL induces apoptosis and NF-kB activation in many tissues and cells.

Specifications

Accession Number P50591
For Use With (Application) Neutralization
Formulation PBS with no preservative; pH 7.4
Gene ID (Entrez) 8743
Molecular Weight (g/mol) 19.6 kDa
Name Human TRAIL (TNFSF10)
pH Range 7.4
Purification Method Purified
Quantity 50 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Gene Alias A330042I21Rik; AI448571; Apo-2 ligand; APO2L; Apo-2L; CD253; CD253 antigen; chemokine tumor necrosis factor ligand superfamily member 10; Ly81; Protein TRAIL; TL2; TN; TNF superfamily member 10; TNF-related apoptosis inducing ligand; TNF-related apoptosis inducing ligand TRAIL; TNF-related apoptosis-inducing ligand; Tnfsf10; TNLG6A; TRAIL; TRAIL protein; TRAIL/APO2L; tumor necrosis factor (ligand) family, member 10; tumor necrosis factor (ligand) superfamily member 10; tumor necrosis factor (ligand) superfamily, member 10; tumor necrosis factor apoptosis-inducing ligand splice variant delta; tumor necrosis factor ligand 6A; tumor necrosis factor ligand superfamily member 10; tumor necrosis factor superfamily member 10
Common Name TRAIL (CD253)
Gene Symbol TNFSF10
Product Type Protein
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Sequence Human TRAIL (TNFSF10) recombinant protein comprises the full-length of the extracellular domain of human TRAIL
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade >97%
Show More Show Less