missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CCL22 (MDC) Recombinant Protein DFS Item

Recombinant Protein

Supplier:  Gibco™ PHC1204

Catalog No. PHC1204


Only null left
Explore available promotions

Description

Description

Carrier-Free Reconstitution: The vial should be centrifuged prior to opening to bring contents to the bottom. The lyophilized powder should be reconstituted with sterile, distilled water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: The lyophilized protein is stable at room temperature for 1 month, but should be kept at -20°C for long term storage. Working aliquots stored with a carrier protein are stable for at least 3 months at -20°C to -80°C. Avoid repeated freeze/thaw cycles. Avoid repeated freeze/thaw cycles.

CCL22 may play a role in the trafficking of activated/effector T-lymphocytes to inflammatory sites and other aspects of activated T-lymphocyte physiology. Also, it is a chemotactic for monocytes. dendritic cells and natural killer cells. CCL22 is a mild chemoattractant for primary activated T-lymphocytes and a potent chemoattractant for chronically activated T-lymphocytes but has no chemattractant activity for neutrophils, eosinophils, and resting T-lymphocytes. Binds to CCR4.
Specifications

Specifications

O00626
Protein with no preservative
8 kDa
Purified
-20°C
<0.1 ng/μg
MDC
ED50 = 10-100 ng/mL; determined by the dose-dependent chemotaxis of human T-cells.
Unconjugated
Recombinant
-20°C
ED50 = 10-100 ng/mL; determined by the dose-dependent chemotaxis of human T-cells.
Approved for shipment on Wet or Dry Ice
Chemokines & Receptors
Lyophilized
Inflammation
>98% by SDS-PAGE
Bioactivity
6367
Human CCL22 (MDC)
20 μg
RUO
A-152E5.1; Abcd1; ABCD-1; Activated B and dendritic cell-derived; AW060611; CC chemokine ABCD-1; CC chemokine STCP-1; C-C motif chemokine 22; C-C motif chemokine ligand 22; Ccl22; chemokine (C-C motif) ligand 22; DC/B-CK; DCBCK; dendritic cell and B cell derived chemokine; macrophage-derived chemokine; MDC; MDC(1-69); MDC(3-69); MDC(5-69); MDC(7-69); MGC108943; MGC34554; RP24-69A20.1; Scya22; small inducible cytokine A22; SMALL INDUCIBLE CYTOKINE A22 PRECURSOR (CC CHEMOKINE ABCD-1) (ACTIVATED B AND DENDRITIC CELL-DERIVED); small inducible cytokine subfamily A (Cys-Cys) member 22; small inducible cytokine subfamily A (Cys-Cys), member 22; small inducible cytokine subfamily A, member 22; small inducible cytokine subfamily A22; small-inducible cytokine A22; STCP-1; stimulated T cell chemotactic protein 1; stimulated T-cell chemotactic protein 1
CCL22
Protein
Human
Human CCL22 recombinant protein contains 67 amino acids
Carrier-Free
<0.1 ng/μg
E. coli
Recombinant, Ligand
Other Proteins
Gibco
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions