missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CCL2 (MCP-1) Recombinant Protein DFS Item

Recombinant Protein

$360.00 - $1424.00

Specifications

Accession Number P13500
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 6347
Molecular Weight (g/mol) 8 kDa
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PHC1011 DFS Item
View Documents
Gibco™
PHC1011
100 μg
Each for $1,424.00
Only null left
 
PHC1014 DFS Item
View Documents
Gibco™
PHC1014
10 μg
Each for $360.00
Only null left
 
PHC1015 DFS Item
View Documents
Gibco™
PHC1015
25 μg
Each for $760.00
Only null left
 
Description

Description

Carrier-Free Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in sterile, distilled water to 0.1-0.5 mg/mL. Apportion the reconstituted protein into working aliquots and store at ≤-20°C. Make any further dilutions of the reconstituted protein in medium or buffered solution containing carrier protein, such as PBS with 0.1% BSA. Storage: Store the lyophilized protein at 2-8°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at ≤-20°C (not in a frost-free freezer). Avoid repeated freeze-thaw cycle.

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.
Specifications

Specifications

P13500
Protein with no preservative
8 kDa
Purified
RUO
Acidic seminal fluid protein; AI323594; C-C motif chemokine 2; C-C motif chemokine ligand 2; Ccl2; chemokine (C-C motif) ligand 2; GDCF-2; HC11; H-MCP-1; HSMCR30; immediate-early serum-responsive JE protein; immediate-early serum-responsive protein JE; Je; MCAF; Mcp1; MCP-1; MCP1A; MCP-1A; MGC9434; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; monocyte chemotactic protein 1A; Monocyte secretory protein JE; monocyte-chemoattractant protein-1 precursor; platelet-derived growth factor-inducible protein JE; RP23-350G1.3; SCYA2; Sigje; small inducible cytokine A2; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; small inducible gene JE; small-inducible cytokine A2; SMC-CF
Ccl2
Protein
Human
4°C
ED50 ≤ 10 ng/mL; determined by the dose-dependent chemotaxis of human THP-1 cells.
Wet Ice
Chemokines & Receptors
Lyophilized
Signal Transduction, Inflammation, Bone Research
≥98% by SDS-PAGE
Bioactivity
6347
Human CCL2 (MCP-1)
4°C
≤0.1 ng/μg
CCL2 (MCP-1)
ED50 ≤ 10 ng/mL; determined by the dose-dependent chemotaxis of human THP-1 cells.
Unconjugated
Recombinant
Carrier-Free
<0.1 ng/μg
E. coli
Recombinant, Ligand
MCP⁄MCAF Chemokines
Gibco
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'