missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CCL17 (TARC) Recombinant Protein DFS Item

Recombinant Protein

Supplier:  Gibco™ PHC1264

Catalog No. PHC1264


Only null left
Explore available promotions

Description

Description

Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute this lyophilized preparation with sterile, distilled water to a concentration of 0.1-1.0 mg/mL. Further dilutions should be made in tissue culture grade buffered solutions with heat inactivated fetal calf serum or low endotoxin BSA. Storage: This lyophilized preparation is stable at 2°C to 8°C, but should be kept at -20°C for long term storage, preferably desiccated. Upon reconstitution, the preparation is stable for up to one week at 2°C to 8°C. For maximal stability, apportion the reconstituted preparation into working aliquots and store at -20°C. Avoid repeated freeze/thaw cycles.

TARC ligand (CCL17) belongs to the CC chemokine family. It is a small cytokine and the gene is located on q arm of chromosome 16, in humans, along with other chemokines. TARC specifically binds to T-cells and interacts with chemokine receptors CCR4 and CCR8. It plays an important role in T cell development in thymus as well as in trafficking and activation of mature T cells.
Specifications

Specifications

Q92583
Protein with no preservative
8 kDa
Purified
-20°C
<0.1 ng/μg
TARC
ED50 = 1 - 10 ng/mL; determined by the dose-dependent chemotaxis of human T-cells.
Unconjugated
Recombinant
-20°C
ED50 = 1 - 10 ng/mL; determined by the dose-dependent chemotaxis of human T-cells.
Approved for shipment on Wet or Dry Ice
Chemokines & Receptors
Lyophilized
Signal Transduction, Immunology
>98% by SDS-PAGE
Bioactivity
6361
Human CCL17 (TARC)
20 μg
RUO
A-152E5.3; ABCD-2; CC chemokine TARC; C-C motif chemokine; C-C motif chemokine 17; C-C motif chemokine ligand 17; CCL17; chemokine (C-C motif) ligand 17; SCYA17; Scya17l; small inducible cytokine subfamily A (Cys-Cys), member 17; small inducible cytokine subfamily A17; small-inducible cytokine A17; T cell-directed CC chemokine; TARC; Thymus and activation-regulated chemokine
CCL17
Protein
Human
Human CCL17 recombinant protein contains 71 amino acids
Carrier-Free
<0.1 ng/μg
E. coli
Recombinant, Ligand
Other Proteins
Gibco
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions