missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human CCL13 (MCP-4) Recombinant Protein DFS Item

Recombinant Protein

Supplier:  Gibco™ PHC1154

Catalog No. PHC1154

Only null left
Explore available promotions

Description

Description

Carrier-Free Reconstitution: We recommend that the vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute this protein in sterile, distilled water to a concentration of 0.1-1.0 mg/mL. Further dilutions should be made in tissue culture grade buffered solutions with heat inactivated fetal calf serum or low endotoxin BSA. Storage: Store lyophilized at ≤+4°C, preferably desiccated. Store reconstituted MCP-4 at -20°C in working aliquots. Avoid repeated freeze-thaw cycles.

Monocyte Chemotactic Protein 4 (MCP-4), also called CCL13, is induced by inflammatory proteins such as IL-1 and TNFalpha. MCP-4 is a ligand for three different G protein coupled receptors, CCR2, CCR3 and CCR5. MCP-4 activates signaling in monocytes, T lymphocytes, eosinophils and basophils and this signaling is associated with the allergic response.
Specifications

Specifications

Q99616
Protein with no preservative
8.6 kDa
Purified
4°C
<0.1 ng/μg
MCP-4
ED50 = 10-100 ng/mL; determined by the dose-dependent chemotaxis of human blood cells.
Unconjugated
Recombinant
4°C
ED50 = 10-100 ng/mL; determined by the dose-dependent chemotaxis of human blood cells.
Approved for shipment on Wet or Dry Ice
Chemokines & Receptors
Lyophilized
Signal Transduction, Immunology
>98% by SDS-PAGE
Bioactivity
6357
Human CCL13 (MCP-4)
20 μg
RUO
C-C motif chemokine 13; C-C motif chemokine 13, long chain; C-C motif chemokine 13, medium chain; C-C motif chemokine 13, short chain; C-C motif chemokine ligand 13; CCL 13; CCL13; chemokine (C-C motif) ligand 13; CKb10; CK-beta-10; H-MCP-4; MCP4; MCP-4; Monocyte chemoattractant protein 4; monocyte chemotactic protein 4; NCC1; NCC-1; new CC chemokine 1; SCYA13; SCYL1; small inducible cytokine subfamily A (Cys-Cys), member 13; small-inducible cytokine A13
CCL13
Protein
Human
Human CCL13 recombinant protein contains 75 amino acids
Carrier-Free
<0.1 ng/μg
E. coli
Recombinant, Ligand
MCP⁄MCAF Chemokines
Gibco
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions