missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human CCDC97 (aa 13-82) Control Fragment Recombinant Protein

Catalog No. RP99112
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99112 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99112 Supplier Invitrogen™ Supplier No. RP99112
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (64%), Rat (64%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60322 (PA5-60322. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

CCDC97 is a coiled-coil domain containing protein.

Specifications

Accession Number Q96F63
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 90324
Name Human CCDC97 (aa 13-82) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CCDC97; coiled-coil domain containing 97; coiled-coil domain-containing protein 97
Common Name CCDC97
Gene Symbol CCDC97
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PDKGCIEPGPGHWGELSRTPVPSKPQDKVEAAEATPVALDSDTSGAENAAVSAMLHAVAASRLPVCSQQQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less