missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CCDC147 (aa 124-209) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP109271
Description
Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-140138 (PA5-140138. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
CFAP58 is a protein coding gene.Specifications
| Q5T655 | |
| Blocking Assay, Control | |
| 159686 | |
| 100 μL | |
| bA127L20.4; bA127L20.4 (novel protein); bA127L20.5; bA127L20.5 (novel protein); bA554P13.1; C10orf80; CCDC147; Cfap58; cilia and flagella associated protein 58; cilia- and flagella-associated protein 58; coiled-coil domain containing 147; coiled-coil domain-containing protein 147; coiled-coil domain-containing protein 147-like; Gm969 | |
| CFAP58 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human CCDC147 (aa 124-209) Control Fragment | |
| RUO | |
| CCDC147 | |
| Unconjugated | |
| Recombinant | |
| KEEIVNLTKLVEQGSGLSMDQHSNIRDLLRFKEEVTKERDQLLSEVVKLRESLAQTTEQQQETERSKEEAEHAISQFQQEIQQRQN | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |