missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Cathepsin Z (aa 71-141) Control Fragment Recombinant Protein

Catalog No. RP103159
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP103159 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP103159 Supplier Invitrogen™ Supplier No. RP103159
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62797 (PA5-62797. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The protein encoded by this gene is a lysosomal cysteine proteinase and member of the peptidase C1 family. It exhibits both carboxy-monopeptidase and carboxy-dipeptidase activities. The encoded protein has also been known as cathepsin X and cathepsin P. This gene is expressed ubiquitously in cancer cell lines and primary tumors and, like other members of this family, may be involved in tumorigenesis.

Specifications

Accession Number Q9UBR2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 1522
Name Human Cathepsin Z (aa 71-141) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI787083; AU019819; carboxypeptidase LB; cathepsin B2; cathepsin IV; cathepsin P; Cathepsin X; cathepsin Y; cathepsin Z; cathepsin Z1; CTSX; CTSZ; cysteine-type carboxypeptidase; D2Wsu143e; lysosomal carboxypeptidase B; preprocathepsin P
Common Name Cathepsin Z
Gene Symbol Ctsz
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence VDGVNYASITRNQHIPQYCGSCWAHASTSAMADRINIKRKGAWPSTLLSVQNVIDCGNAGSCEGGNDLSVW
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less