missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Cathepsin H ELISA Kit

Catalog No. EH75RBX5
Encompass
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH75RBX5 5 x 96 Tests
EH75RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH75RBX5 Supplier Invitrogen™ Supplier No. EH75RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions

ELISA

Human Cathepsin H quantitates human Cathepsin H in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cathepsin H.

The cathepsin family of proteolytic enzymes contains several diverse classes of proteases. The cysteine protease class comprises cathepsins B, L, H, K, S, and O. The aspartyl protease class is composed of cathepsins D and E. Cathepsin G is in the serine protease class. Most cathepsins are lysosomal and each is involved in various cellular events such as peptide biosynthesis and protein degradation. The propeptide region of cathepsin H is highly homologous to other cathepsins. This region is thought to play a role in the processing of cysteine proteases. Cathepsin H is thought to be involved in several pathological conditions, including neuro degenerative disorders and cancer.

Specifications

Accession Number P09668
Assay Range 0.17-40 ng/mL
Assay Sensitivity 0.17 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias ACC4; ACC-4; ACC5; ACC-5; AL022844; aleurain; Cat H; CATH; cathepsin B3; Cathepsin BA; cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; CPSB; Ctsh; N-benzoylarginine-beta-naphthylamide hydrolase; pro-cathepsin HACC4; ACC-4; ACC5; ACC-5; AL022844; aleurain; Cat H; CATH; cathepsin B3; Cathepsin BA; cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; CPSB; Ctsh; N-benzoylarginine-beta-naphthylamide hydrolase; pro-cathepsin H;
Gene ID (Entrez) 1512
Gene Symbol CTSH
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Cathepsin H
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less