Learn More
Gibco™ Human Cardiotrophin-1 Recombinant Protein
Recombinant Protein
Supplier: Gibco™ PHC1594
Description
Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute Cardiotrophin-1 in 20 mM Tris, pH 8.0, to a concentration of 0.1-1.0 mg/mL. Further dilutions should be made in aqueous buffers supplemented with carrier protein, such as 0.1-1.0% BSA. Storage: This lyophilized preparation is stable at 2°C to 8°C, but should be kept at -20°C for long term storage, preferably desiccated. Upon reconstitution, the preparation is stable for up to one week at 2°C to 8°C. For maximal stability, apportion the reconstituted preparation into working aliquots and store at -20°C. Avoid repeated freeze/thaw cycles.
Cardiotrophin-1 (CT-1) is a member of the IL-6 family of cytokines, which signal through gp130 receptor complexes. gp130 complexes with several different receptor subunits to transmit signals from Cardiotrophin-1, IL-6, LIF, OSM, CNTF and IL-11. Cardiotrophin-1 binds to and activates the leukemia inhibitory factor (LIF) receptor/gp130 receptor complex and has been shown to induce hypertrophy in cardiac myocytes in vitro. Cardiotrophin-1, a secreted protein expressed at high levels in myocardium during cardiogenesis, has been shown to promote proliferation and survival of embryonic cardiomyocytes, suggesting a role for Cardiotrophin-1 in the activation of gp130 during cardiac development.Specifications
| Q16619 | |
| Protein with no preservative | |
| 21.5 kDa | |
| Purified | |
| -20°C | |
| <0.1 ng/μg | |
| Cardiotrophin 1 | |
| ED50 ≤ 1.0 ng/mL; determined by the dose-dependent proliferation of TF-1 cells. | |
| Unconjugated | |
| Recombinant | |
| -20°C | |
| ED50 ≤ 1.0 ng/mL; determined by the dose-dependent proliferation of TF-1 cells. | |
| Approved for shipment on Wet or Dry Ice | |
| Cytokines & Receptors | |
| Lyophilized | |
| Neurobiology, Stem Cell Research, Cardiovascular Research, Angiogenesis | |
| ≥98% by SDS-PAGE and HPLC |
| Bioactivity | |
| 1489 | |
| Human Cardiotrophin-1 | |
| 10 μg | |
| RUO | |
| cardiophin 1; Cardiotrophin; cardiotrophin 1; Cardiotrophin1; cardiotrophin-1; CT 1; CT1; CT-1; CT-1CT1; CTF 1; Ctf1; IL-6 cytokine family | |
| CTF1 | |
| Protein | |
| Human | |
| Human Cardiotrophin-1 recombinant protein contains 200 amino acids | |
| Carrier-Free | |
| <0.1 ng/μg | |
| E. coli | |
| Recombinant, Ligand | |
| Other Proteins | |
| Gibco |