missing translation for 'onlineSavingsMsg'
Learn More

Human Calreticulin-2 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Catalog No. IC2927G10
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC2927G10 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IC2927G10 Supplier Bio-Techne Supplier No. IC2927G100UG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Calreticulin-2/CALR3 Monoclonal specifically detects Calreticulin-2/CALR3 in Human samples. It is validated for Flow Cytometry (Intracellular Staining).

Specifications

Antigen Calreticulin-2/CALR3
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 321007
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q96L12
Gene Alias CALR3, calreticulin 2, calreticulin 3, Calreticulin2, Calreticulin-2, calreticulin-3, cancer/testis antigen 93, Crt2, CRT2calreticulin-2, CT93, FLJ25355, MGC26577
Gene Symbols CALR3
Host Species Mouse
Immunogen E. coli-derived recombinant human Calreticulin-2, Thr20-Leu384, Accession # Q96L12
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 125972
Test Specificity Detects human Calreticulin-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Calreticulin is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Product Type Antibody
Form Supernatant
Isotype IgG2b
Product Line R&D Systems
Show More Show Less